missing translation for 'onlineSavingsMsg'
Learn More

RAP1A Antibody (RAP1 4C8/1) - BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody

€ 190.00 - € 351.00

Specifications

Antigen RAP1A
Clone RAP1 4C8/1
Concentration 0.43 mg/mL
Dilution Western Blot 1:100 - 1:2000, Flow Cytometry 1:10 - 1:1000, ELISA 1:100 - 1:2000, Immunohistochemistry 1:10 - 1:500, Immunocytochemistry/ Immunofluorescence 1:10 - 1:500, CyTOF-ready
Applications Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunofluorescence
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18619656
View Documents
Novus Biologicals
NBP2-50269-0.025ml
0.025 mL
€ 190.00
0.03mL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18677508
View Documents
Novus Biologicals
NBP2-50269
0.1 mL
€ 351.00
0.10mL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

RAP1A Monoclonal antibody specifically detects RAP1A in Human, Mouse samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, CyTOF
Specifications

Specifications

RAP1A
0.43 mg/mL
Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunofluorescence
Unconjugated
Mouse
Cell Cycle and Replication, Endosome Markers
PBS
C21KG, G-22K, GTP-binding protein smg p21A, KREV-1, KREV1Ras-related protein Krev-1, RAP1, RAP1A, member of RAS oncogene family, ras-related protein Rap-1A, RAS-related protein RAP1A, SMGP21
Human RAP1A
Primary
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
RAP1 4C8/1
Western Blot 1:100 - 1:2000, Flow Cytometry 1:10 - 1:1000, ELISA 1:100 - 1:2000, Immunohistochemistry 1:10 - 1:500, Immunocytochemistry/ Immunofluorescence 1:10 - 1:500, CyTOF-ready
Monoclonal
Purified
RUO
Human, Mouse
A2ALS5
5906
IgG2b
Protein A purified
Videos
SDS
Documenti

Documenti

Certificati
Correzione del contenuto del prodotto

Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.

Titolo del prodotto

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.

Grazie per averci aiutato a migliorare il nostro sito web. Il vostro feedback è stato inviato