missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ RAP1GAP Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5106715
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total RAP1GAP.
Rap1GAP specifically stimulates GTP hydrolytic activity of the monomeric G protein Rap1. Physical interaction between G alpha z, a member of the Gi family of trimeric G proteins, and Rap1GAP blocks the ability of regulators of G protein signaling to stimulate GTP hydrolysis of the alpha subunit, and also attenuates the ability of activated G alpha z to inhibit adenylyl cyclase. Rap1GAP is expressed in the brain, kidney and pancreas and may act as a signal integrator to coordinate and/or integrate Gz signaling and Rap1 signaling in cells. A novel isoform of Rapl GTPase-activating protein, designated Rap1GAPII, binds specifically to G alpha z. Stimulation of the Gi-coupled M2 Muscarinic receptor translocates Rap1GAPII from the cytosol to the membrane and decreases the amount of GTP-bound Rap1, resulting in the activation of ERK/MAPK.
Specifications
| RAP1GAP | |
| Polyclonal | |
| Unconjugated | |
| RAP1GAP | |
| 1300019I11Rik; 2310004O14Rik; AI427470; ARPP-90; KIAA0474; RAP1 GTPase activating protein; Rap1 GTPase-activating protein; rap1 GTPase-activating protein 1; Rap1, GTPase-activating protein 1; Rap1ga1; Rap1GAP; Rap1GAP1; RAP1GAPII; RAPGAP | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 110351, 313644, 5909 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| A2ALS5, P47736 | |
| RAP1GAP | |
| A synthesized peptide derived from human RAP1GAP(Accession P47736), corresponding to amino acid residues S484-S534. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido