missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RAP1GAP2 Rabbit Polyclonal Antibody, Proteintech
Rabbit Polyclonal Antibody
€ 145.00 - € 410.00
Specifications
| Antigen | RAP1GAP2 |
|---|---|
| Concentration | 1500 μg/mL |
| Applications | Western Blot, Immunohistochemistry, ELISA |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30237236
|
Proteintech
25391-1-AP-20UL |
20 μL |
€ 145.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30236455
|
Proteintech
25391-1-AP-150UL |
150 μL |
€ 410.00
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The RAP1GAP2 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human RAP1GAP2. This antibody recognizes human, mouse, rat antigen. The RAP1GAP2 antibody has been validated for the following applications: WB, IHC, ELISA analysis.Specifications
| RAP1GAP2 | |
| Western Blot, Immunohistochemistry, ELISA | |
| Unconjugated | |
| Rabbit | |
| Human, Mouse, Rat | |
| DKFZp686O238; KIAA1039; RAP1GA3; Rap1GAP2 | |
| IgG | |
| Antigen affinity purification | |
| Polyclonal Antibody |
| 1500 μg/mL | |
| Polyclonal | |
| Liquid | |
| RUO | |
| 23108 | |
| Recombinant Protein | |
| Primary | |
| Store at -20°C. Stable for one year after shipment. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title