missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Rap2A Antibody, Allophycocyanin, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-24574APC
This item is not returnable.
View return policy
Description
Rap2A Polyclonal antibody specifically detects Rap2A in Human, Mouse, Canine, Chicken, Equine, Opossum, Primate, Xenopus samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
| Rap2A | |
| Polyclonal | |
| Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin | |
| KREV, K-REV, RAP2, RAP2, member of RAS oncogene family (K-rev), RAP2A, member of RAS oncogene family, ras-related protein Rap-2a, RbBP-30 | |
| A portion of amino acids 140-183 of human RAP2A was used as the immunogen for the antibody. | |
| 0.1 mL | |
| Signal Transduction | |
| 5911 | |
| Store at 4C in the dark. | |
| IgG |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| APC | |
| PBS | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Canine, Chicken, Equine | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction