missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Rap2B Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-20075
This item is not returnable.
View return policy
Description
Rap2B Polyclonal specifically detects Rap2B in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| Rap2B | |
| Polyclonal | |
| Unconjugated | |
| 0.1M Tris (pH 7.0), 0.1M Glycine and 20% Glycerol with 0.01% Thimerosal | |
| MGC20484, RAP2B, member of RAS oncogene family, Ras family small GTP binding protein RAP2B, ras-related protein Rap-2b, small GTP binding protein | |
| Rabbit | |
| 21 kDa | |
| 0.1 mL | |
| Signal Transduction | |
| 5912 | |
| Human, Mouse | |
| IgG |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| 0.97mg/mL | |
| Western Blot 1:500-1:3000, Immunohistochemistry 1:100-1:1000, Immunohistochemistry-Paraffin 1:100-1:1000 | |
| P61225 | |
| RAP2B | |
| Recombinant protein encompassing a sequence within the center region of human RAP2B. The exact sequence is proprietary. | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Human, Mouse. Species reactivity based on sequence identity: Rhesus Monkey(100%), Rat (100%). | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction