missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RAP80 Antibody, DyLight 755, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB100-2346IR
This item is not returnable.
View return policy
Description
RAP80 Polyclonal antibody specifically detects RAP80 in Human samples. It is validated for Western Blot
Specifications
| RAP80 | |
| Polyclonal | |
| Western Blot | |
| BRCA1-A complex subunit RAP80, RAP80X2HRIP110, receptor associated protein 80, Receptor-associated protein 80, retinoid x receptor interacting protein, Retinoid X receptor-interacting protein 110, RXRIP110, ubiquitin interaction motif containing 1, Ubiquitin interaction motif-containing protein 1 | |
| The immunogen recognized by this antibody maps to a region between residue 600 and 650 of human Receptor Associated Protein 80 using the numbering given in entry NP_057374.3 (GeneID 51720). | |
| 0.1 mL | |
| Breast Cancer, Cancer | |
| 51720 | |
| Store at 4C in the dark. | |
| IgG |
| Western Blot | |
| DyLight 755 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction