missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
RAP80 Polyclonal antibody specifically detects RAP80 in Human,Mouse,Rat samples. It is validated for ELISA,Western Blot,Immunocytochemistry/Immunofluorescence
Specifications
Specifications
| Antigen | RAP80 |
| Applications | ELISA, Western Blot, Immunocytochemistry/Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1:500 - 1:2000, ELISA, Immunocytochemistry/Immunofluorescence 1:50 - 1:100 |
| Formulation | PBS (pH 7.3), 50% glycerol |
| Gene Alias | BRCA1-A complex subunit RAP80, RAP80X2HRIP110, receptor associated protein 80, Receptor-associated protein 80, retinoid x receptor interacting protein, Retinoid X receptor-interacting protein 110, RXRIP110, ubiquitin interaction motif containing 1, Ubiquitin interaction motif-containing protein 1 |
| Host Species | Rabbit |
| Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 1-150 of human RAP80 (NP_057374.3).,, Sequence:, MPRRKKKVKEVSESRNLEKKDVETTSSVSVKRKRRLEDAFIVISDSDGEEPKEENGLQKTKTKQSNRAKCLAKRKIAQMTEEEQFALALKMSEQEAREVNSQEEEEEELLRKAIAESLNSCRPSDASATRSRPLATGPSSQSHQEKTTDS |
| Purification Method | Affinity purified |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?