missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RAP80 Antibody (SR2242), Novus Biologicals™
Rabbit Monoclonal Antibody
€ 338.00 - € 449.00
Specifications
| Antigen | RAP80 |
|---|---|
| Clone | SR2242 |
| Dilution | Western Blot 1:1000-1:5000, Immunohistochemistry 1:50-1:200, Immunocytochemistry/Immunofluorescence 1:50-1:200 |
| Applications | Western Blot, Immunohistochemistry, Immunofluorescence |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18629923
|
Novus Biologicals
NBP3-21834-100ul |
100 μg |
€ 449.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18660884
|
Novus Biologicals
NBP3-21834-50ul |
50 μg |
€ 338.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
RAP80 Monoclonal antibody specifically detects RAP80 in Human samples. It is validated for Western Blot, Immunohistochemistry, ImmunofluorescenceSpecifications
| RAP80 | |
| Western Blot 1:1000-1:5000, Immunohistochemistry 1:50-1:200, Immunocytochemistry/Immunofluorescence 1:50-1:200 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| BRCA1-A complex subunit RAP80, RAP80X2HRIP110, receptor associated protein 80, Receptor-associated protein 80, retinoid x receptor interacting protein, Retinoid X receptor-interacting protein 110, RXRIP110, ubiquitin interaction motif containing 1, Ubiquitin interaction motif-containing protein 1 | |
| A synthesized peptide derived from human RAP80 (Uniprot #: Q96RL1) | |
| Primary | |
| Store at -20° C. Avoid freeze/thaw cycles. |
| SR2242 | |
| Western Blot, Immunohistochemistry, Immunofluorescence | |
| Unconjugated | |
| Rabbit | |
| Breast Cancer, Cancer | |
| PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
| 51720 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title