missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RAPGEF1 Monoclonal Antibody (OTI2C3), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA506838
This item is not returnable.
View return policy
Description
This gene encodes a human guanine nucleotide exchange factor. It transduces signals from CRK by binding the SH3 domain of CRK, and activating several members of the Ras family of GTPases. This signaling cascade that may be involved in apoptosis, integrin-mediated signal transduction, and cell transformation. Several alternatively spliced transcript variants of this gene have been described, but the full-length nature of some variants has not been determined.Specifications
| RAPGEF1 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q13905 | |
| Rapgef1 | |
| Full length human recombit protein of human RAPGEF1 produced in HEK293T cell. | |
| 100 μL | |
| Primary | |
| Canine, Human, Mouse, Rat | |
| Antibody | |
| IgG1 |
| Immunohistochemistry (Paraffin), Western Blot | |
| OTI2C3 | |
| Unconjugated | |
| RAPGEF1 | |
| C3G, GRF2 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 107746, 2889, 491286, 63881 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction