Learn More
Thermo Scientific™ RapID™ YEAST PLUS System
Identify over 40 different yeasts in just four hours using this rapid identifcation system based on enzyme technology.
Brand: Thermo Scientific™ R8311007
328.70 EUR valid until 2024-05-31
Use promo code "21664" to get your promotional price.
Additional Details : Weight : 0.01000kg Transport : UN number : 1814 Chem class : 8 Pack group : II
Description
Identify over 40 different yeasts in just four hours using the Thermo Scientific™ RapID™ YEAST PLUS System, an identification system based on enzyme technology.
The RapID YEAST PLUS System is a qualitative micromethod employing conventional and chromogenic substrates for the identification of medically important yeast, yeast-like and related organisms which have been isolated from human clinical specimens.- One-step inoculation—decreased prep time for increased productivity
- Manual microbial identification made easy—no oil, no pipetting, no 24-hr incubation
- Four-hour incubation*—quicker time-to-result for a faster response
- Common procedure—lessened materials usage saves time and money
- Visible color reactions—reduced subjectivity decreases repeat testing
- Better reporting—more coverage, advanced updates
*Two-hour incubation for RapID SS/u.
A clear plastic tray contains reagent impregnated wells, allowing simultaneuos inoculation of each cavity with a predetermined volume of inoculum. A suspension of test organism in RapID Innoculation Fluid is used as the inoculum which rehydrates and initiates test reactions.
After incubation, each cavity is examined for reactivity by noting the development of a color. In some cases, reagents must be added to the cavities to provide a color change. The resulting pattern of positive and negative scores is used as the basis for identification of the test isolate by comparison of test results to reactivity patterns stored in a databse or through the use of a computer-generated Code Compedium (ERIC).
Requires:
RapID Inoculation Fluid (R8325106)
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand, combining powerful manual, semi-automated and fully automated test products and a comprehensive line of media and diagnostic products to offer a complete, end-to-end solution to quickly deliver the products you need and the quality results your laboratory depends on.
Specifications
2°C to 8°C | |
Yes | |
1 Box |
RapID YEAST PLUS System | |
Yeasts | |
20 Panels/Kit |
Safety and Handling
- Danger
Hazard Category
- Reproductive toxicity Category 1B
Hazard Statement
- H360Df-May damage the unborn child. Suspected of damaging fertility.
Precautionary Statement
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.