missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ RAPSN Polyclonal Antibody

Rabbit Polyclonal Antibody

Brand:  Invitrogen™ PA593169

Product Code. 16528411

  • € 530.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Immunogen sequence: MGQDQTKQQI EKGLQLYQSN QTEKALQVWT KVLEKSSDLM GRFRVLGCLV TAHSEMGRYK EMLKFAVVQI DTARELEDAD FLLESYLNLA RSNEKLCEFH KTISYCKTCL GLPGTRAGAQ LGGQVSLSMG NAFLGLSVFQ KALESFEKAL RYAHNNDDAM LECRVCCSLG SFYAQVKDYE KALFFPCKAA ELVNNYGKGW SLKYRAMSQY HMAVAYRLLG RLGSAMECCE ESMKIALQHG DRPLQALCLL CFADIHRSRG DLELSQLKLH CLSESIYRSK GLQRELRAHV VRFHECVEET ELYCGLCGES IGEKNSRLQA LPCSHIFHLR CLQNNGTRSC PNCRRSSMKP GFV; Positive Samples: B-cell, Mouse skeletal muscle, Mouse brain, Rat skeletal muscle, Rat brain; Cellular Location: Cell junction, Cell membrane, Cytoplasm, Cytoplasmic side, Peripheral membrane protein, cytoskeleton, postsynaptic cell membrane, synapse.

This gene encodes a member of a family of proteins that are receptor associated proteins of the synapse. The encoded protein contains a conserved cAMP-dependent protein kinase phosphorylation site, and plays a critical role in clustering and anchoring nicotinic acetylcholine receptors at synaptic sites by linking the receptors to the underlying postsynaptic cytoskeleton, possibly by direct association with actin or spectrin. Mutations in this gene may play a role in postsynaptic congenital myasthenic syndromes. Alternatively spliced transcript variants encoding multiple isoforms have been observed for this gene.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

RAPSN
Polyclonal
Unconjugated
RAPSN
43 kD receptor-associated protein of the synapse; 43 kda postsynaptic protein; 43 kDa receptor-associated protein of the synapse; 43kDa; 43kDa acetylcholine receptor-associated protein; acetylcholine receptor-associated 43 kda protein; AChR associated protein at synapses; acetylcholine receptor associated protein at synapses; 43kDa protein; likely to cluster nicotinic AChRs; CMS11; CMS1D; CMS1E; CMS4C; FADS; MGC3597; Nraps; Raps; Rapsn; rapsyn; receptor associated protein of the synapse; receptor-associated protein of the synapse; receptor-associated protein of the synapse, 43 kDa; receptor-associated protein of the synapse, 43kD; RING finger protein 205; RNF205; twitch once; two
Rabbit
Affinity Chromatography
RUO
19400, 362161, 5913
-20°C, Avoid Freeze/Thaw Cycles
Liquid
ELISA, Immunohistochemistry (Paraffin), Western Blot
0.3 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
P12672, Q13702
RAPSN
Recombinant protein (or fragment).
100 μL
Primary
Human, Mouse, Rat
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.