missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RAR alpha/NR1B1 Antibody (OTI2C3), Janelia Fluor™ 646, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-73802JF646
This item is not returnable.
View return policy
Description
RAR alpha/NR1B1 Monoclonal specifically detects RAR alpha/NR1B1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.
Specifications
| RAR alpha/NR1B1 | |
| Monoclonal | |
| Janelia Fluor 646 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| RARA | |
| Recombinant protein fragment corresponding to amino acids 52-299 of human RAR alpha/NR1B1 (NP_000955) produced in E.coli. | |
| 0.1 mL | |
| Primary | |
| Human, Mouse, Rat | |
| IgG1 |
| Western Blot, Immunohistochemistry | |
| OTI2C3 | |
| Western Blot, Immunohistochemistry | |
| alpha polypeptide, RAR-alpha, retinoic acid receptor, alpha | |
| Mouse | |
| Affinity Purified | |
| Cancer, Phospho Specific, Vision | |
| 5914.0 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction