missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RAR gamma/NR1B3 Antibody (SR1891), Novus Biologicals™
Rabbit Monoclonal Antibody
€ 338.00 - € 449.00
Specifications
| Antigen | RAR gamma/NR1B3 |
|---|---|
| Clone | SR1891 |
| Dilution | Western Blot 1:500-1:2000, Immunocytochemistry/Immunofluorescence 1:50-1:200 |
| Applications | Western Blot, Immunofluorescence |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18639674
|
Novus Biologicals
NBP3-21974-100ul |
100 μg |
€ 449.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18649684
|
Novus Biologicals
NBP3-21974-50ul |
50 μg |
€ 338.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
RAR gamma/NR1B3 Monoclonal antibody specifically detects RAR gamma/NR1B3 in Human, Rat samples. It is validated for Western Blot, ImmunofluorescenceSpecifications
| RAR gamma/NR1B3 | |
| Western Blot 1:500-1:2000, Immunocytochemistry/Immunofluorescence 1:50-1:200 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Rat | |
| NR1B3RARC, Nuclear receptor subfamily 1 group B member 3, RAR-gamma, retinoic acid receptor gamma, retinoic acid receptor, gamma | |
| A synthesized peptide derived from human RAR gamma/NR1B3 (Uniprot #: P13631) | |
| Primary | |
| Store at -20° C. Avoid freeze/thaw cycles. |
| SR1891 | |
| Western Blot, Immunofluorescence | |
| Unconjugated | |
| Rabbit | |
| Cancer, Transcription Factors and Regulators | |
| PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
| 5916 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title