missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RARA Monoclonal Antibody (OTI7B9), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA806341S
This item is not returnable.
View return policy
Description
RARA Monoclonal antibody specifically detects RARA in Human samples. It is validated for Western BlotSpecifications
| RARA | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| P10276 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 5914 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG1 |
| Western Blot | |
| OTI7B9 | |
| Unconjugated | |
| RARA | |
| Nr1b1; nuclear receptor subfamily 1 group B member 1; nucleophosmin-retinoic acid receptor alpha fusion protein NPM-RAR long form; RAR; RAR Alpha; RAR alpha 1; RARA; RAR-alpha; RARalpha1; retinoic acid; retinoic acid nuclear receptor alpha variant 1; retinoic acid nuclear receptor alpha variant 2; retinoic acid receptor; retinoic acid receptor alpha; retinoic acid receptor, alpha; retinoic acid receptor, alpha polypeptide; RRA | |
| Human recombinant protein fragment corresponding to amino acids 52-299 of human RARA produced in E.coli. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction