missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ RARA/RARB Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5115840
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total Retinoic Acid Receptor alpha/beta.
Retinoic acid, a metabolite of vitamin A, is necessary for normal organogenesis but acts as a teratogen at high levels during embryonic and fetal development. Retinoic acid functions through its interaction with the nuclear protein, retinoic acid receptor (RAR). RAR belongs to the steroid and thyroid hormone superfamily of nuclear receptor proteins which exert their effects by binding to specific DNA response elements, thus regulating gene expression in target cells. RAR exists as three major subtypes: alpha, beta and gamma.
Specifications
| RARA/RARB | |
| Polyclonal | |
| Unconjugated | |
| RARA | |
| A830025K23; HAP; HBV-activated protein; hepatitis B virus activated protein; MCOPS12; Nr1b1; Nr1b2; Nuclear receptor subfamily 1 group B member 1; nuclear receptor subfamily 1 group B member 2; nucleophosmin-retinoic acid receptor alpha fusion protein NPM-RAR long form; RAR; RAR alpha 1; RARA; RAR-alpha; RARalpha1; Rarb; RAR-beta; RARbeta1; RAR-epsilon; retinoic acid; retinoic acid nuclear receptor alpha variant 1; retinoic acid nuclear receptor alpha variant 2; retinoic acid receptor; retinoic acid receptor alpha; Retinoic acid receptor beta; retinoic acid receptor, alpha; retinoic acid receptor, alpha polypeptide; retinoic acid receptor, beta; retinoic acid receptor, beta polypeptide; RRB2 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 19401, 218772, 24705, 24706, 5914, 5915 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide | |
| P10276, P10826, P11416, P22605 | |
| RARA, RARB | |
| A synthesized peptide derived from human RARA(Accession P10276), corresponding to amino acid residues P73-T123. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction