missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RARRES1 Antibody (OTI1D2), Biotin, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-73803B
This item is not returnable.
View return policy
Description
RARRES1 Monoclonal specifically detects RARRES1 in Human, Mouse, Rat, Canine, Monkey samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.
Specifications
| RARRES1 | |
| Monoclonal | |
| Biotin | |
| PBS with 0.05% Sodium Azide | |
| RARRES1 | |
| Full length recombinant protein of human RARRES1 (NP_996846) produced in HEK293T cells. | |
| 0.1 mL | |
| Primary | |
| Human, Mouse, Rat, Canine, Monkey | |
| IgG2b |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence | |
| OTI1D2 | |
| Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence | |
| RAR-responsive protein TIG1, retinoic acid receptor responder (tazarotene induced) 1, retinoic acid receptor responder protein 1, TIG1Tazarotene-induced gene 1 protein | |
| Mouse | |
| Affinity Purified | |
| Cell Cycle and Replication | |
| 5918.0 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction