missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RARRES1 Monoclonal Antibody (OTI1B1), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA506084
This item is not returnable.
View return policy
Description
This gene was identified as a retinoid acid (RA) receptor-responsive gene. It encodes a type 1 membrane protein. The expression of this gene is upregulated by tazarotene as well as by retinoic acid receptors. The expression of this gene is found to be downregulated in prostate cancer, which is caused by the methylation of its promoter and CpG island. Alternatively spliced transcript variant encoding distinct isoforms have been observed.Specifications
| RARRES1 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| P49788 | |
| RARRES1 | |
| Full length human recombit protein of human RARRES1 produced in HEK293T cell. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG2b |
| Immunohistochemistry (Paraffin), Western Blot | |
| OTI1B1 | |
| Unconjugated | |
| RARRES1 | |
| LXNL, PERG-1, TIG1 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 5918 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction