missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ RARRES3 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA597156
This item is not returnable.
View return policy
Description
Immunogen sequence: MASPHQEPKP GDLIEIFRLG YEHWALYIGD GYVIHLAPPS EYPGAGSSSV FSVLSNSAEV KRERLEDVVG GCCYRVNNSL DHEYQPRPVE VIISSAKEMV GQKMKYSIVS RNCEHFVTQL RYGKSRCKQV EKAKV; Positive Samples: 293T; Cellular Location: Membrane, Single-pass membrane protein.
Retinoids exert biologic effects such as potent growth inhibitory and cell differentiation activities and are used in the treatment of hyperproliferative dermatological diseases. These effects are mediated by specific nuclear receptor proteins that are members of the steroid and thyroid hormone receptor superfamily of transcriptional regulators. RARRES1, RARRES2, and RARRES3 are genes whose expression is upregulated by the synthetic retinoid tazarotene. RARRES3 is thought act as a tumor suppressor or growth regulator.
Specifications
| RARRES3 | |
| Polyclonal | |
| Unconjugated | |
| PLAAT4 | |
| HRAS-like suppressor 4; HRASLS4; HRSL4; Phospholipase A and acyltransferase 4; PLA1/2-3; PLAAT4; RARRES3; RAR-responsive protein TIG3; retinoic acid receptor responder (tazarotene induced) 3; retinoic acid receptor responder 3; retinoic acid receptor responder protein 3; retinoic acid-inducible gene 1; retinoid-inducible gene 1 protein; RIG1; Tazarotene-induced gene 3 protein; TIG3 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 5920 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot | |
| 0.52 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q9UL19 | |
| PLAAT4 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-135 of human RARRES3 (NP_0045762). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction