Learn More
Invitrogen™ RARS Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA552035
Description
Immunogen sequence: TIIGESISRL FEFAGYDVLR LNHVGDWGTQ FGMLIAHLQD KFPDYLTVSP PIGDLQVFYK ESKKRFDTEE EFKKRAYQCV VLLQGKNPDI TKAWKLICDV SRQELNKIYD ALDVSLIERG ESFYQDRMND IVKEFEDRGF VQVDDGRKIV Highest antigen sequence identity to the following orthologs: Mouse - 91%, Rat - 91%.
The fidelity of protein synthesis requires efficient discrimination of amino acid substrates by aminoacyl-tRNA synthetases. ArgRS (Arginyl-tRNA synthetase), also known as RARS or DALRD1, belongs to the class-I aminoacyl-tRNA synthetase family that includes the related proteins, LeuRS, ValRS and IleRS. These proteins are large monomeric proteins and play a major role in catalyzing the aminoacylation of tRNA by their cognate amino acid. ArgRS localizes to the cytoplasm and exists as a monomer but can also associate with other tRNA synthetases and auxiliary proteins to form a multisubunit complex. In the presence of ATP, arginine (Arg) and tRNA, ArgRS joins Arg to tRNA(Arg) at its synthetic active site. Two cytoplasmic forms of ArgRS have been described in mammals, differing by the addition of a 73 amino acid sequence that is required for ArgRS assembly into the multisubunit complex.
Specifications
| RARS | |
| Polyclonal | |
| Unconjugated | |
| RARS1 | |
| 2610011N19Rik; 2610037E21Rik; AL033339; arginine tRNA ligase 1, cytoplasmic; arginine--tRNA ligase, cytoplasmic; Arginyl-tRNA synthetase; arginyl-tRNA synthetase, cytoplasmic; argRS; AW985894; DALRD1; HLD9; MGC8641; RARS | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 5917 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| P54136 | |
| RARS1 | |
| Recombinant protein corresponding to Human RARS. Recombinant protein control fragment (Product #RP-102263). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.