missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ras-GAP Antibody (OTI1H2), DyLight 488, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-73804G
This item is not returnable.
View return policy
Description
Ras-GAP Monoclonal specifically detects Ras-GAP in Human, Mouse, Rat samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.
Specifications
| Ras-GAP | |
| Monoclonal | |
| DyLight 488 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| Mouse | |
| Affinity Purified | |
| Cancer, Core ESC Like Genes, Neuronal Cell Markers, Neuroscience, Phospho Specific, Signal Transduction, Stem Cell Markers | |
| 5921.0 | |
| Store at 4°C in the dark. |
| Western Blot, Immunocytochemistry, Immunofluorescence | |
| OTI1H2 | |
| Western Blot, Immunocytochemistry/Immunofluorescence | |
| RASA1 | |
| Full length recombinant protein of human RAS p21 (NP_002881) produced in HEK293T cells. | |
| 0.1 mL | |
| Primary | |
| Human, Mouse, Rat | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction