missing translation for 'onlineSavingsMsg'
Learn More

RASA1 Monoclonal Antibody (OTI1A4), TrueMAB™, OriGene

Product Code. 17594992
Encompass_Preferred
Change view
Click to view available options
Quantity:
30 μL
100 μL
Unit Size:
100µL
30µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17594992 100 μL 100µL
17515002 30 μL 30µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17594992 Supplier Origene Technologies Supplier No. TA505901

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

GTPase Activating Protein (GAP) is capable of stimulating the hydrolysis of GTP by ras p21 proteins, though GAP has little effect on the oncogenic forms of ras. It is also known that several tyrosine kinases such as platelet derived growth factor receptor and epidermal growth factor receptor are involved in the tyrosine phosphorylation of GAP. It has therefore been suggested that GAP may provide a connection or link between growth factor receptors and the ras p21 family.

Specifications

Antigen RASA1
Applications Western Blot
Classification Monoclonal
Clone OTI1A4
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene RASA1
Gene Accession No. P20936
Gene Alias CM-AVM, CMAVM, GAP, PKWS, RASA, RASGAP, p120, p120GAP, p120RASGAP
Gene Symbols RASA1
Host Species Mouse
Immunogen Full length human recombit protein of human RASA1 produced in HKE293T cell.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5921
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.