Learn More
RASA4 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand:  Novus Biologicals NBP2-15089
Product Code. 18179081
  • ONLINE EXCLUSIVE PRICE
    € 629.00
    € 594.30 / 0.10mL
    Save € 34.70
    5% Off
Description
RASA4 Polyclonal specifically detects RASA4 in Human samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
RASA4
Polyclonal
Unconjugated
0.1M Tris (pH 7.0), 0.1M Glycine and 20% Glycerol with 0.01% Thimerosal
RASA4
Recombinant protein encompassing a sequence within the N-terminus region of human RASA4. The exact sequence is proprietary.
0.1 mL
Primary
Human
IgG
Western Blot, Immunocytochemistry, Immunofluorescence
0.9mg/mL
Western Blot 1:500-1:3000, Immunocytochemistry/Immunofluorescence 1:100-1:1000
Ca2+-promoted Ras inactivator, Calcium-promoted Ras inactivator, CAPRIGAPLKIAA0538RASA4B, FLJ59070, Putative Ras GTPase-activating protein 4B, ras GTPase-activating protein 4, RAS p21 protein activator 4MGC131890, RasGAP-activating-like protein 2
Rabbit
Affinity Purified
RUO
10156
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.