missing translation for 'onlineSavingsMsg'
Learn More

RASAL3 Polyclonal Antibody
GREENER_CHOICE

Product Code. 13218158
Change view
Click to view available options
Quantity:
400 μL
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
13218158 400 μL 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 13218158 Supplier Invitrogen Supplier No. PA524097

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

RASAL3 Polyclonal Antibody

RASAL3 Polyclonal Antibody for Western Blot, IHC (P)

TRUSTED_SUSTAINABILITY

Specifications

Antigen RASAL3
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene RASAL3
Gene Accession No. Q86YV0
Gene Alias RAS protein activator like 3; RAS protein activator like-3; RASAL3
Gene Symbols RASAL3
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide between 947-975 amino acids from the C-terminal region of human RASAL3.
Purification Method Antigen affinity chromatography, Protein A
Quantity 400 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 64926
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.