missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RASD2 Antibody (OTI1F7), PE/Cy7, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-73808PECY7
This item is not returnable.
View return policy
Description
RASD2 Monoclonal specifically detects RASD2 in Human, Mouse, Rat samples. It is validated for Flow Cytometry.
Specifications
| RASD2 | |
| Monoclonal | |
| PE-Cyanine7 | |
| PBS with 0.05% Sodium Azide | |
| RASD2 | |
| Full length recombinant protein of human RASD2 (NP_055125) produced in HEK293T cells. | |
| 0.1 mL | |
| Primary | |
| Human, Mouse, Rat | |
| IgG1 |
| Flow Cytometry | |
| OTI1F7 | |
| Flow Cytometry | |
| GTP-binding protein Rhes, MGC:4834, RASD family, member 2, Rhes, TEM2Ras homolog enriched in striatum, Tumor endothelial marker 2 | |
| Mouse | |
| Affinity Purified | |
| Signal Transduction | |
| 23551.0 | |
| Store at 4°C in the dark. Do not freeze. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction