Learn More
RASEF Antibody (S03-1G6), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand:  Novus Biologicals NBP3-19968-50ul
 View more versions of this product
Product Code. 18660525
  • € 421.00 / 50µL
Description
RASEF Monoclonal antibody specifically detects RASEF in Human, Mouse, Rat samples. It is validated for Western Blot, Immunocytochemistry/ Immunofluorescence
TRUSTED_SUSTAINABILITY
Specifications
Specifications
RASEF
Monoclonal
0.3 mg/mL
50mM Tris-Glycine(pH 7.4), 0.15M NaCl, 40% Glycerol, 0.05% BSA
Rabbit
Affinity purified
RUO
Primary
Human, Mouse, Rat
Purified
Western Blot, Immunofluorescence
S03-1G6
Unconjugated
FLJ31614, RAB45, RAB45, member RAS oncogene family, RAS and EF hand domain containing, RAS and EF-hand domain containing, ras and EF-hand domain-containing protein, Ras-related protein Rab-45
A synthetic peptide of human RASEF (Uniprot # Q8IZ41)
50 μg
Signal Transduction
158158
Store at 4°C short term. Store at -20°C long term. Avoid freeze-thaw cycles.