missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ RASGEF1A Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA557329
This item is not returnable.
View return policy
Description
Immunogen sequence: HIELDRVSSI YPEDLMQIVS HMDSLDNHRC RGDLTKTYSL EAYDNWFNCL SMLVATEVCR VVKKKHRTRM Highest antigen sequence identity to the following orthologs: Mouse - 91%, Rat - 93%.
Guanine nucleotide exchange factor (GEF) with specificity for RAP2A, KRAS, HRAS, and NRAS (in vitro). Plays a role in cell migration.
Specifications
| RASGEF1A | |
| Polyclonal | |
| Unconjugated | |
| RASGEF1A | |
| 6330404M18Rik; AI835194; CG4853; CG4853 gene product; RasGEF domain family member 1A; RasGEF domain family, member 1A; ras-GEF domain-containing family member 1A; Rasgef1a | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 221002 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.3 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q8N9B8 | |
| RASGEF1A | |
| Recombinant protein corresponding to Human RASGEF1A. Recombinant protein control fragment (Product #RP-97216). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction