missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RASGRF2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-20087
This item is not returnable.
View return policy
Description
RASGRF2 Polyclonal specifically detects RASGRF2 in Mouse, Rat samples. It is validated for Western Blot.
Specifications
| RASGRF2 | |
| Polyclonal | |
| Unconjugated | |
| 0.1M Tris (pH 7.0), 0.1M Glycine and 10% Glycerol with 0.01% Thimerosal | |
| DKFZp781H1715, GRF2RAS-GRF2, guanine nucleotide-releasing factor 2, Ras guanine nucleotide exchange factor 2, Ras protein-specific guanine nucleotide-releasing factor 2, Ras-GRF2, ras-specific guanine nucleotide-releasing factor 2 | |
| Rabbit | |
| 141 kDa | |
| 0.1 mL | |
| Primary | |
| Mouse. Species reactivity based on sequence identity: Human (100%). | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
| Western Blot | |
| 1mg/mL | |
| Western Blot 1:500-1:3000 | |
| O14827 | |
| RASGRF2 | |
| Recombinant protein encompassing a sequence within the C-terminus region of human RASGRF2. The exact sequence is proprietary. | |
| Affinity Purified | |
| RUO | |
| 5924 | |
| Mouse | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction