missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
Antibody detects endogenous levels of total RASGRF2.
RAS (MIM 190020) GTPases cycle between an inactive GDP-bound state and an active GTP-bound state. Guanine-nucleotide exchange factors (GEFs), such as RASGRFs, stimulate the conversion of the GDP-bound form into the active form.
Specifications
Specifications
| Antigen | RASGRF2 |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.02% sodium azide |
| Gene | RASGRF2 |
| Gene Accession No. | O14827, P70392, Q99JE4 |
| Gene Alias | 6330417G04Rik; AW048350; Grf2; guanine nucleotide-releasing factor 2; ras guanine nucleotide exchange factor 2; Ras protein specific guanine nucleotide releasing factor 2; RAS protein-specific guanine nucleotide-releasing factor 2; Rasgrf2; ras-GRF2; Ras-specific guanine nucleotide-releasing factor 2 |
| Gene Symbols | RASGRF2 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?
