missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ RASGRP2 Polyclonal Antibody
GREENER_CHOICE

Product Code. 15347524
Change view
Click to view available options
Quantity:
400 μL
Unit Size:
400µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15347524 400 μL 400µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15347524 Supplier Invitrogen™ Supplier No. PA535111

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

This antibody is predicted to react with bovine, mouse and rat based on sequence homology.

The protein encoded by this gene is a brain-enriched nucleotide exchanged factor that contains an N-terminal GEF domain, 2 tandem repeats of EF-hand calcium-binding motifs, and a C-terminal diacylglycerol/phorbol ester-binding domain. This protein can activate small GTPases, including RAS and RAP1/RAS3. The nucleotide exchange activity of this protein can be stimulated by calcium and diacylglycerol. Three alternatively spliced transcript variants encoding the same protein have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RASGRP2
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene RASGRP2
Gene Accession No. Q7LDG7
Gene Alias Calcium and DAG-regulated guanine nucleotide exchange factor I; calcium and diacylglycerol-regulated guanine nucleotide exchange factor I; Caldaggef1; CalDAG-GEFI; CDC25L; cdc25-like protein; F25B3.3 kinase like protein; F25B3.3 kinase-like protein; guanine exchange factor MCG7; hCDC25L; MCG7; RAP 1A protein-specific guanine nucleotide exchange factor 1; RAS guanyl nucleotide-releasing protein 2; RAS guanyl releasing protein 2; RAS guanyl releasing protein 2 (calcium and DAG-regulated); RAS guanyl-releasing protein 2; RAS, guanyl releasing protein 2; RAS, guanyl releasing protein 2; RAP 1A protein-specific guanine nucleotide exchange factor 1; CalDAG-GEFI; Rasgrp2
Gene Symbols RASGRP2
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide between 7-34 amino acids from the N-terminal region of human RASGRP2.
Purification Method Antigen affinity chromatography, Protein A
Quantity 400 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10235
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.