missing translation for 'onlineSavingsMsg'
Learn More

RASGRP3 Monoclonal Antibody (OTI3C5), TrueMAB™, OriGene

Product Code. 17559812
Encompass_Preferred
Change view
Click to view available options
Quantity:
30 μL
100 μL
Unit Size:
100µL
30µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17559812 30 μL 30µL
17539812 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17559812 Supplier Origene Technologies Supplier No. TA801445S

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

RASGRP3 Monoclonal antibody specifically detects RASGRP3 in Human samples. It is validated for Western Blot

Specifications

Antigen RASGRP3
Applications Western Blot
Classification Monoclonal
Clone OTI3C5
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene NME1
Gene Accession No. P15531
Gene Alias BC066069; calcium and DAG-regulated guanine nucleotide exchange factor III; calcium- and diacylglycerol-regulated guanine nucleotide exchange factor III; CalDAG-GEFIII; Gm327; GRP3; Guanine nucleotide exchange factor for Rap1; KIAA0846; OTTHUMP00000201245; RAS guanyl releasing protein 3; RAS guanyl releasing protein 3 (calcium and DAG-regulated); ras guanyl-releasing protein 3; RAS, guanyl releasing protein 3; Rasgrp3
Host Species Mouse
Immunogen Full length human recombinant protein of human NME1 produced in E.coli.
Purification Method Affinity chromatography
Quantity 30 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4830
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.