Learn More
RASGRP3 Monoclonal Antibody (OTI3B9), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand:  Origene Technologies TA801566
Encompass_Preferred
 View more versions of this product
Product Code. 17549922
  • € 540.87 / 100µL
Description
Members of the RAS subfamily of GTPases function in signal transduction as GTP/GDP-regulated switches that cycle between inactive GDP- and active GTP-bound states. Guanine nucleotide exchange factors (GEFs), such as RASGRP3, serve as RAS activators by promoting acquisition of GTP to maintain the active GTP-bound state and are the key link between cell surface receptors and RAS activation.
Specifications
Specifications
RASGRP3
Monoclonal
1 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Q8IV61
RASGRP3
Human recombit protein fragment corresponding to amino acids 380-690 of human RASGRP3 produced in E.coli.
100 μL
Primary
Human
Antibody
IgG2b
Western Blot
OTI3B9
Unconjugated
RASGRP3
GRP3
Mouse
Affinity Chromatography
RUO
25780
-20° C, Avoid Freeze/Thaw Cycles