missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Neurturin Antibody (32E21), Alexa Fluor™ 532, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-22024AF532
This item is not returnable.
View return policy
Description
Neurturin Monoclonal specifically detects Neurturin in Human samples. It is validated for Western Blot, ELISA.
Specifications
| Neurturin | |
| Monoclonal | |
| Western Blot, ELISA | |
| neurturin, NTN | |
| Mouse | |
| Protein G purified | |
| Cytokine Research, Immunology | |
| 4902.0 | |
| Store at 4°C in the dark. |
| Western Blot, ELISA | |
| Alexa Fluor 532 | |
| 50 mM sodium borate with 0.05% sodium azide | |
| NRTN | |
| Human recombinant Neurturin | |
| 0.1 mL | |
| Primary | |
| Human | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction