missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RASL11B Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals H00065997-D01P
This item is not returnable.
View return policy
Description
RASL11B Polyclonal antibody specifically detects RASL11B in Human samples. It is validated for Western Blot
Specifications
| RASL11B | |
| Polyclonal | |
| Western Blot 1:500 | |
| NP_076429.1 | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot | |
| Unconjugated | |
| PBS (pH 7.4) | |
| MGC2827, MGC4499, ras-like protein family member 11B, RAS-like, family 11, member B | |
| RASL11B (NP_076429.1, 1 a.a. - 248 a.a.) full-length human protein. MRLIQNMCTIAEYPAPGNAAASDCCVGAAGRRLVKIAVVGASGVGKTALVVRFLTKRFIGDYERNAGNLYTRQVQIEGETLALQVQDTPGIQVHENSLSCSEQLNRCIRWADAVVIVFSITDYKSYELISQLHQHVQQLHLGTRLPVVVVANKADLLHIKQVDPQLGLQLASMLGCSFYEVSVSENYNDVYSAFHVLCKEVSHKQQPSSTPEKRRTSLIPRPKSPNMQDLKRRFKQALSAKVRTVTSV | |
| 0.1 mg | |
| Signal Transduction | |
| 65997 | |
| Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction