missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RASSF1 Antibody (OTI2B11), DyLight 594, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-73786DL594
This item is not returnable.
View return policy
Description
RASSF1 Monoclonal antibody specifically detects RASSF1 in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)
Specifications
| RASSF1 | |
| Monoclonal | |
| DyLight 594 | |
| 123F2, NORE2A, pancreas-specific ras association domain family 1 protein, Ras association (RalGDS/AF-6) domain family member 1, ras association domain-containing protein 1, RASSF1A, RDA32cardiac-specific ras association domain family 1 protein, REH3P21, tumor suppressor protein RDA32, WUGSC:H_LUCA12.5 | |
| Full length recombinant protein of human RASSF1 (NP_009113) produced in E.coli. | |
| 0.1 mL | |
| Breast Cancer, Cell Cycle and Replication, Mitotic Regulators | |
| 11186 | |
| Store at 4°C in the dark. | |
| IgG1 |
| Western Blot, Flow Cytometry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| OTI2B11 | |
| 50mM Sodium Borate | |
| Mouse | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction