missing translation for 'onlineSavingsMsg'
Learn More

RASSF1 Monoclonal Antibody (OTI8E2), TrueMAB™, OriGene

Product Code. p-200011092
Encompass_Preferred
Change view
Click to view available options
Quantity:
30 μL
100 μL
Unit Size:
100µL
30µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17517021 30 μL 30µL
17597011 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17517021 Supplier Origene Technologies Supplier No. TA502486S

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

RASSF1 Monoclonal antibody specifically detects RASSF1 in Human samples. It is validated for Immunocytochemistry, Western Blot

Specifications

Antigen RASSF1
Applications Immunocytochemistry, Western Blot
Classification Monoclonal
Clone OTI8E2
Concentration 0.87 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene HPGD
Gene Accession No. P15428
Gene Alias 123F protein; 123F2; AA536941; AU044980; cardiac-specific ras association domain family 1 protein; NORE2A; pancreas-specific ras association domain family 1 protein; protein 123F2; Ras association (RalGDS/AF-6) domain family 1; Ras association (RalGDS/AF-6) domain family member 1; Ras association domain family 1; Ras association domain family member 1; ras association domain-containing protein 1; Rassf1; Rassf1A; RASSF1A tumor suppressor; Rassf1B; Rassf1C; RDA32; REH3P21; tumor suppressor protein RDA32; wu:fc17f02; WUGSC:H_LUCA12.5; zgc:112224; zgc:92505; zgc:92505 protein
Host Species Mouse
Immunogen Full length human recombinant protein of human HPGD produced in HEK293T cell.
Purification Method Affinity chromatography
Quantity 30 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3248
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Form Liquid
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.