Learn More
RASSF2 Polyclonal Antibody
GREENER_CHOICE
RASSF2 Polyclonal Antibody
Brand:  Invitrogen PA598113
Product Code. 16520942
  • € 473.00 / 100µL
Description
RASSF2 Polyclonal Antibody for Western Blot, IP, ELISA
TRUSTED_SUSTAINABILITY
Specifications
Specifications
RASSF2
Polyclonal
Unconjugated
RASSF2
3830431H01Rik; 9030412M04Rik; AI852669; AW495050; CENP-34; centromere protein 34; Kiaa0168; Ras association (RalGDS/AF-6) domain family 2; Ras association (RalGDS/AF-6) domain family member 2; Ras association domain family member 2; ras association domain-containing protein 2; RASFADIN; Rassf2; RP4-599I11.1
Rabbit
Protein G
RUO
9770
-20°C or -80°C if preferred
Liquid
ELISA, Immunoprecipitation, Western Blot
1.83 mg/mL
PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
P50749
RASSF2
Recombinant Human Ras association domain-containing protein 2 protein (69-326AA).