Learn More
Invitrogen™ RASSF4 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5140202
Description
Gently mix before use. Optimal concentrations and conditions for each application should be determined by the user. Immunogen sequence: DGPSEFALYI VHESGERTKL KDCEYPLISR ILHGPCEKIA RIFLMEADLG V Highest antigen sequence indentity to the following orthologs: Rat - 88%, Mouse - 88%.
The function of this gene has not yet been determined but may involve a role in tumor suppression. Alternative splicing of this gene results in several transcript variants; however, most of the variants have not been fully described.
Specifications
| RASSF4 | |
| Polyclonal | |
| Unconjugated | |
| RASSF4 | |
| 3830411C14Rik; AD037; AI586137; Ras association (RalGDS/AF-6) domain family 4; Ras association (RalGDS/AF-6) domain family member 4; Ras association domain family 4; Ras association domain family member 4; ras association domain-containing protein 4; Rassf4; tumor suppressor RASSF4 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 83937 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunocytochemistry | |
| 0.05 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9H2L5 | |
| RASSF4 | |
| Recombinant protein corresponding to Human RASSF4. Recombinant protein control fragment (Product #RP-109609). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.