missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RASSF6 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-93092-0.1ml
This item is not returnable.
View return policy
Description
RASSF6 Polyclonal antibody specifically detects RASSF6 in Human, Mouse samples. It is validated for Western Blot
Specifications
| RASSF6 | |
| Polyclonal | |
| Western Blot 1:500-1:2000 | |
| DKFZp686K23225, putative RAS binding protein, Ras association (RalGDS/AF-6) domain family 6, Ras association (RalGDS/AF-6) domain family member 6, ras association domain-containing protein 6 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-303 of human RASSF6 (NP_001257320.1). MTMMAHQYPSWIFINEKTFITREQLNSLLKTYNIFYENQKNLHILYGETEDGKLIVEGMLDIFWGVKRPIQLKIQDEKPFSSFTSMKSSDVFSSKGMTRWGEFDDLYRISELDRTQIPMSEKRNSQEDYLSYHSNTLKPHAKDEPDSPVLYRTMSEAALVRKRMKPLMMDRKERQKNRASINGHFYNHEIENSPQDFALHIIFATGEQRRLKKTDIPLLQRLLQGPSEKNARIFLMDKDAEEISSDVAQYINFHFSLLESILQRLNEEEKREIQRIVTKFNKEKAIILKCLQNKLVIKTETTV | |
| 0.1 mL | |
| Apoptosis, Cancer, Cell Biology, GPCR, Signal Transduction | |
| 166824 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction