missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ RASSF6 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA576480
This item is not returnable.
View return policy
Description
The antibody was affinity-purified from rabbit antiserum by affinity-chromatography using epitope-specific immunogen and the purity is > 95% (by SDS-PAGE).
RASSF6 is a member of the RAS association domain family of RAS effectors/tumor suppressors that mediate some of the growth inhibitory properties of RAS oncogenes.
Specifications
| RASSF6 | |
| Polyclonal | |
| Unconjugated | |
| Rassf6 | |
| 1600016B17Rik; DKFZp686K23225; putative RAS binding protein; Ras association (RalGDS/AF-6) domain family 6; Ras association (RalGDS/AF-6) domain family member 6; Ras association domain family member 6; ras association domain-containing protein 6; RASSF6 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 166824, 305251, 73246 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q4QR82, Q6ZTQ3, Q80UQ2 | |
| Rassf6 | |
| Recombinant fusion protein of human RASSF6. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction