Learn More
RASSF8 Mouse anti-Human, Clone: OTI1F12, lyophilized, TrueMAB™
Mouse Monoclonal Antibody
Brand: Origene Technologies CF505923
Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
RASSF8 encodes a member of the Ras-association domain family (RASSF) of tumor suppressor proteins. This gene is essential for maintaining adherens junction function in epithelial cells and has a role in epithelial cell migration. It is a lung tumor suppressor gene candidate. A chromosomal translocation t(12;22)(p11.2;q13.3) leading to the fusion of this gene and the FBLN1 gene is found in a complex type of synpolydactyly. Multiple alternatively spliced transcript variants encoding different isoforms have been found for this gene.Specifications
| RASSF8 | |
| Monoclonal | |
| Unconjugated | |
| RASSF8 | |
| C12orf2, HOJ1 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 11228 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Lyophilized |
| Immunocytochemistry, Immunofluorescence, Western Blot | |
| OTI1F12 | |
| PBS with 8% trehalose and no preservative; pH 7.3 | |
| Q8NHQ8 | |
| RASSF8 | |
| Full length human recombit protein of human RASSF8 produced in HEK293T cell. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.