missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ RASSF8 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5104058
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total RASSF8.
RASSF8 encodes a member of the Ras-association domain family (RASSF) of tumor suppressor proteins. This gene is essential for maintaining adherens junction function in epithelial cells and has a role in epithelial cell migration. It is a lung tumor suppressor gene candidate. A chromosomal translocation t(12;22)(p11.2;q13.3) leading to the fusion of this gene and the FBLN1 gene is found in a complex type of synpolydactyly. Multiple alternatively spliced transcript variants encoding different isoforms have been found for this gene.
Specifications
| RASSF8 | |
| Polyclonal | |
| Unconjugated | |
| RASSF8 | |
| 5133400D11Rik; AA815821; AW123240; C12orf2; Carcinoma-associated protein HOJ-1; carcinoma-associated protein HOJ-1 homolog; EGK_03473; HOJ1; HoJ-1; mHoj-1; Ras association (RalGDS/AF-6) domain family (N-terminal) member 8; Ras association (RalGDS/AF-6) domain family 8; Ras association domain family member 8; ras association domain-containing protein 8; RASSF8; RGD1308820 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 11228, 312846, 71323 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q8CJ96, Q8NHQ8 | |
| RASSF8 | |
| A synthesized peptide derived from human RASSF8(Accession Q8NHQ8), corresponding to amino acid residues S385-V419. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction