missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RASSF9 Monoclonal Antibody (OTI5E5), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA809393S
This item is not returnable.
View return policy
Description
RASSF9 Monoclonal antibody specifically detects RASSF9 in Human samples. It is validated for Western BlotSpecifications
| RASSF9 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q9C0B1 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 79068 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG1 |
| Western Blot | |
| OTI5E5 | |
| Unconjugated | |
| Fto | |
| AW322107; PAM COOH-terminal interactor protein 1; Pamci; PCIP1; P-cip1; Peptidylglycine alpha-amidating monooxygenase COOH-terminal interactor; peptidylglycine alpha-amidating monooxygenase COOH-terminal interactor protein-1; Ras association (RalGDS/AF-6) domain family (N-terminal) member 9; Ras association domain family member 9; ras association domain-containing protein 9; RASSF9 | |
| Full length human recombinant protein of human FTO produced in HEK293T cell. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction