missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Rat Albumin Polyclonal Antibody, FITC
GREENER_CHOICE

Product Code. 11563791
Change view
Click to view available options
Quantity:
1 mg
Unit Size:
1mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11563791 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11563791 Supplier Invitrogen™ Supplier No. PA186695

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Sheep Polyclonal Antibody

Sheep anti Rat albumin antibody recognizes rat albumin, a soluble protein which constitutes approximately 50% of blood serum protein, acting as a regulator of blood colloidal osmotic pressure and hence blood volume.

Albumin is a soluble protein which constitutes approximately 50% of blood serum protein, and acts as a regulator of blood colloidal osmotic pressure and blood volume. Albumin is also an important transporter of substances within the bloodstream, such as fatty acids, thyroid hormones, metal ions and steroids.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Rat Albumin
Applications ELISA, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate FITC
Formulation PBS with 0.09% sodium azide
Gene Alb
Gene Accession No. P02769, P02770
Gene Alias Alb; ALB protein; Alb1; albumin; Albza; Bos d 6; bovine serum albumin; BSA; Serum albumin
Gene Symbols Alb
Host Species Sheep
Immunogen Native protein.
Purification Method Protein G
Quantity 1 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 24186, 280717
Target Species Rat, Bovine
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. Store in the dark.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.