missing translation for 'onlineSavingsMsg'
Learn More

ABCC4, Rat, Clone: M4I-10, Abnova™

Rat monoclonal antibody raised against recombinant ABCC4.

Brand:  Abnova MAB3400.1mL

Product Code. 16045806

  • € 1250.00 / 1mL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

The protein encoded by this gene is a member of the superfamily of ATP-binding cassette (ABC) transporters. ABC proteins transport various molecules across extra- and intra-cellular membranes. ABC genes are divided into seven distinct subfamilies (ABC1, MDR/TAP, MRP, ALD, OABP, GCN20, White). This protein is a member of the MRP subfamily which is involved in multi-drug resistance. The specific function of this protein has not yet been determined; however, this protein may play a role in cellular detoxification as a pump for its substrate, organic anions. Alternative splicing results in multiple splice variants encoding different isoforms. [provided by RefSeq

Specifications

Specifications

ABCC4
Monoclonal
Unconjugated
Immunohistochemistry (Frozen sections) (1:20) Immunocytochemistry (1:20-1:50) Western Blot (1:20-1:50) The optimal working dilution should be determined by the end user.
ABCC4
ABCC4
Recombinant protein corresponding to chimeric protein with amino acids 372-431 of human ABCC4 and bacterial maltose-binding protein.
RUO
10257
Human, Mouse
Liquid
Flow Cytometry, Immunocytochemistry, Immunohistochemistry (Frozen), Western Blot
M4I-10
Rat monoclonal antibody raised against recombinant ABCC4.
In serum-free culture supernatant (0.09% sodium azide)
EST170205/MOAT-B/MOATB/MRP4
Rat
1 mL
Primary
MRP4 transports cyclic nucleotides and anti-retroviral compounds. Anti-MRP4 clone M4I-10 reacts with human multidrug resistance protein 4 as well as with the mouse orthologue of the transporter molecule (Mrp4).
Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
IgG2a
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.