Learn More
BCL2L2, Rat, Clone: 13F9, Abnova™
Rat monoclonal antibody raised against full-length recombinant BCL2L2.
Brand: Abnova MAB3487.100ug
Description
This gene encodes a member of the BCL-2 protein family. The proteins of this family form hetero- or homodimers and act as anti- and pro-apoptotic regulators. Expression of this gene in cells has been shown to contribute to reduced cell apoptosis under cytotoxic conditions. Studies of the related gene in mice indicated a role in the survival of NGF- and BDNF-dependent neurons. Mutation and knockout studies of the mouse gene demonstrated an essential role in adult spermatogenesis. [provided by RefSeq
Specifications
| BCL2L2 | |
| Monoclonal | |
| Unconjugated | |
| The optimal working dilution should be determined by the end user. | |
| BCL2L2 | |
| BCL2L2 | |
| Recombinant His fusion protein corresponding to full length human BCL2L2. | |
| RUO | |
| 599 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG2a |
| Immunoprecipitation | |
| 13F9 | |
| Rat monoclonal antibody raised against full length recombinant BCL2L2. | |
| In PBS (0.02% sodium azide) | |
| BCL-W/BCLW/KIAA0271 | |
| Rat | |
| 100 μg | |
| Primary | |
| Canine, Human, Monkey, Mouse, Rat | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.