missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cd4, Rat, PE-TXRD, Clone: GK1.5, Abnova™
Rat monoclonal antibody raised against Cd4.
Brand: Abnova MAB5920.100ug
This item is not returnable.
View return policy
Specifications
| Cd4 | |
| Monoclonal | |
| PE-Texas Red | |
| Flow Cytometry (0.2 ug/106 cells) The optimal working dilution should be determined by the end user. | |
| Cd4 | |
| Cd4 | |
| Cloned mouse CTL line V41. | |
| RUO | |
| 12504 | |
| Mouse | |
| Liquid |
| Flow Cytometry, Immunohistochemistry (Frozen), Immunoprecipitation | |
| GK1.5 | |
| Rat monoclonal antibody raised against Cd4. | |
| In PBS (0.09% sodium azide) | |
| L3T4/Ly-4 | |
| Rat | |
| 100 μg | |
| Primary | |
| CD4/L3T4a, Mr 52 KDa. | |
| Store in the dark at 4°C. Do not freeze. Avoid prolonged exposure to light. Aliquot to avoid repeated freezing and thawing. |
|
| IgG2b κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction