missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cd80, Rat, SPRD, Clone: 1G10, Abnova™
Rat monoclonal antibody raised against native Cd80.
Brand: Abnova MAB5952.100ug
This item is not returnable.
View return policy
Specifications
| Cd80 | |
| Monoclonal | |
| SPRD | |
| Flow Cytometry (0.2 ug/106 cells) The optimal working dilution should be determined by the end user. | |
| Cd80 | |
| Cd80 | |
| Native purified Cd80 from dibutyryl cAMP-activated 5C2 cells. | |
| RUO | |
| 12519 | |
| Mouse | |
| Liquid |
| Flow Cytometry, Immunoprecipitation | |
| 1G10 | |
| Rat monoclonal antibody raised against native Cd80. | |
| In PBS (0.09% sodium azide) | |
| B7-1/B7.1/Cd28l/Ly-53/Ly53/MIC17/TS/A-1 | |
| Rat | |
| 100 μg | |
| Primary | |
| Specificity Mouse CD80 (B7-1), Mr 45-47 KDa | |
| Store in the dark at 4°C. Do not freeze. Avoid prolonged exposure to light. Aliquot to avoid repeated freezing and thawing. |
|
| IgG2a κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction