missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cd86, Rat, PE-Cy7, Clone: GL1, Abnova™
Rat monoclonal antibody raised against native Cd86.
Brand: Abnova MAB5756.100ug
This item is not returnable.
View return policy
Specifications
| Cd86 | |
| Monoclonal | |
| PE-Cyanine7 | |
| Flow Cytometry (0.1 ug/106 cells) The optimal working dilution should be determined by the end user. | |
| Cd86 | |
| Cd86 | |
| Native purified Cd86 from LPS-activated mouse B cells. | |
| RUO | |
| 12524 | |
| Mouse | |
| Liquid |
| Flow Cytometry, Immunohistochemistry (Frozen), Immunoprecipitation | |
| GL1 | |
| Rat monoclonal antibody raised against native Cd86. | |
| In PBS (0.09% sodium azide) | |
| B7/B7-2/B7.2/B70/CLS1/Cd28l2/ETC-1/Ly-58/Ly58/MB7/MB7-2/TS/A-2 | |
| Rat | |
| 100 μg | |
| Primary | |
| Mouse CD86, the B7-2 co-stimulatory molecule and a ligand for CD28 and CD152/CTLA-4. | |
| Store in the dark at 4°C. Do not freeze. Avoid prolonged exposure to light. Aliquot to avoid repeated freezing and thawing. |
|
| IgG2a κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction