missing translation for 'onlineSavingsMsg'
Learn More

Cd9, Rat, FITC, Clone: EM-04, Abnova™

Product Code. 16066686
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16066686 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16066686 Supplier Abnova Supplier No. MAB5120.100ug

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rat monoclonal antibody raised against native Cd9.

Specifications

Antigen Cd9
Applications Flow Cytometry
Classification Monoclonal
Clone EM-04
Concentration 0.5 mg/mL
Conjugate FITC
Description Rat monoclonal antibody raised against native Cd9.
Dilution Flow Cytometry (6 ug/mL) The optimal working dilution should be determined by the end user.
Formulation In PBS (0.2% BSA, 0.09% sodium azide)
Gene Cd9
Gene Alias Tspan29
Gene Symbols Cd9
Host Species Rat
Immunogen Native purified Cd9 from permeabilized mouse bone marrow-derived mast cells (BMMC).
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 12527
Test Specificity This antibody recognizes CD9 antigen, a 24 KDa transmembrane protein expressed on platelets, monocytes, pre-B lymphocytes, granulocytes and activated T lymphocytes.
Target Species Mouse
Content And Storage Store in the dark at 4°C. Do not freeze.
Avoid prolonged exposure to light.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.