missing translation for 'onlineSavingsMsg'
Learn More

Csf2, Rat, Biotin, Clone: MP1-31G6, Abnova™

Product Code. 16231210
Change view
Click to view available options
Quantity:
500 μg
Unit Size:
500µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16231210 500 μg 500µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16231210 Supplier Abnova Supplier No. MAB5886.500ug

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rat monoclonal antibody raised against recombinant Csf2.

Specifications

Antigen Csf2
Applications ELISA
Classification Monoclonal
Clone MP1-31G6
Conjugate Biotin
Description Rat monoclonal antibody raised against recombinant Csf2.
Dilution The optimal working dilution should be determined by the end user.
Formulation In PBS (0.09% sodium azide)
Gene Csf2
Gene Alias Csfgm/Gm-CSf/MGC151255/MGC151257/MGI-IGM
Gene Symbols Csf2
Host Species Rat
Immunogen Recombinant protein corresponding to mouse Csf2.
Quantity 500 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 12981
Test Specificity Mouse Granulocyte/Macrophage Colony Stimulating Factor (GM-CSF)1-3. This is a neutralizing antibody.
Target Species Mouse
Content And Storage Store at 4°C.
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.